Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ LSMEM1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA565488
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA565488 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA565488 Supplier Invitrogen™ Supplier No. PA565488
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MDDVSRRLTAE GKDIDDLKRI NNMIVKRLNQ LNQLDSEQN Highest antigen sequence identity to the following orthologs - mouse 88%, rat 85%.

LSMEM1 is a protein coding gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LSMEM1
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene LSMEM1
Gene Accession No. Q8N8F7
Gene Alias C7orf53; Gm889; leucine rich single-pass membrane protein 1; leucine-rich single-pass membrane protein 1; Lsmem1
Gene Symbols LSMEM1
Host Species Rabbit
Immunogen Recombinant Human LSMEM1. Recombinant protein control fragment (Product #RP-106162).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 286006
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.