Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ LSMEM2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. FEPA5144155
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEPA5144155 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FEPA5144155 Supplier Invitrogen™ Supplier No. PA5144155
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

LSMEM2 Polyclonal Antibody for ELISA, ICC/IF, IHC (P)

LSMEM2 (Leucine Rich Single-Pass Membrane Protein 2) is a Protein Coding gene. Predicted to be integral component of membrane.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LSMEM2
Applications ELISA, Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% Proclin 300; pH 7.4
Gene LSMEM2
Gene Accession No. Q8N112
Gene Alias C3orf45Leucine-rich single-pass membrane protein 2; LSMEM2
Gene Symbols LSMEM2
Host Species Rabbit
Immunogen Recombinant Human Leucine-rich single-pass membrane protein 2 protein (1-96AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 132228
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.