Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
LSP1 Antibody (LSP1/3042), DyLight 550, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP308286R
Description
LSP1 Monoclonal antibody specifically detects LSP1 in Human samples. It is validated for ELISA, Protein ArraySpecifications
LSP1 | |
Monoclonal | |
DyLight 550 | |
47 kDa actin binding protein, 47 kDa actin-binding protein, 52 kDa phosphoprotein, F-actin binding and cytoskeleton associated protein, leufactin (leukocyte F-actin binding protein), leukocyte-specific protein 1, lymphocyte-specific protein 1, lymphocyte-specific protein pp52, pp52, WP34Lymphocyte-specific antigen WP34 | |
Recombinant fragment of human LSP1 protein (around aa174-278) (exact sequence is proprietary) (Uniprot: P33241) | |
100 μg | |
Primary | |
Human | |
Purified |
ELISA, Protein Array | |
LSP1/3042 | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified | |
RUO | |
4046 | |
Store at 4°C in the dark. | |
IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction