Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ LSP1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA582689
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA582689 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA582689 Supplier Invitrogen™ Supplier No. PA582689
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: PRTPSPLVLE GTIEQSSPPL SPTTKLIDRT ESLNRSIEKS NSVKKSQPDL PISKIDQWLE QYTQAIETAG RTPKLARQAS IELPSMAVAS TK Highest antigen sequence indentity to the following orthologs: Rat - 82%, Mouse - 82%.

This gene encodes an intracellular F-actin binding protein. The protein is expressed in lymphocytes, neutrophils, macrophages, and endothelium and may regulate neutrophil motility, adhesion to fibrinogen matrix proteins, and transendothelial migration. Alternative splicing results in multiple transcript variants encoding different isoforms.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LSP1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene LSP1
Gene Accession No. P33241
Gene Alias 1 BAC CH230-300K22 (Children's Hospital Oakland Research Institute) complete; 47 kDa actin binding protein; 47 kDa actin-binding protein; 52 kDa phosphoprotein; F-actin binding and cytoskeleton associated protein; leufactin (leukocyte F-actin binding protein); leukocyte specific protein 1; leukocyte-specific protein 1; lsp; LSP1; Lsp-1; lymphocyte specific 1; lymphocyte specific protein 1; lymphocyte-specific antigen WP34; Lymphocyte-specific protein 1; OTTHUMP00000014138; OTTHUMP00000069614; OTTHUMP00000069615; OTTHUMP00000069616; p50; pp52; S37; S37 protein; WP34
Gene Symbols LSP1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human LSP1. Recombinant protein control fragment (Product #RP-92040).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4046
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.