Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
LSP1 Rabbit anti-Human, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
Supplier: Proteintech Group Inc 155931AP150UL
Description
This gene encodes an intracellular F-actin binding protein. The protein is expressed in lymphocytes, neutrophils, macrophages, and endothelium and may regulate neutrophil motility, adhesion to fibrinogen matrix proteins, and transendothelial migration. Alternative splicing results in multiple transcript variants encoding different isoforms.Specifications
| LSP1 | |
| Polyclonal | |
| Unconjugated | |
| LSP1 | |
| 47 kDa actin binding protein, 52 kDa phosphoprotein, LSP1, lymphocyte protein 1, Lymphocyte specific protein 1, pp52, WP34 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 4046 | |
| -20°C | |
| Liquid |
| Immunoprecipitation, Western Blot | |
| 0.42 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P33241 | |
| LSP1 | |
| LSP1 Fusion Protein Ag7944 | |
| 150 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction