Learn More
LSR Rabbit anti-Human, Polyclonal, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A304470A

Description
The recommended shelf life for this product is 1 year from date of receipt.
Probable role in the clearance of triglyceride-rich lipoprotein from blood. Binds chylomicrons, LDL and VLDL in presence of free fatty acids and allows their subsequent uptake in the cells.Specifications
LSR | |
Polyclonal | |
Unconjugated | |
LSR | |
ILDR3, immunoglobulin-like domain containing receptor 3, lipolysis-stimulated remt, LISCH protein, LISCH7, liver-specific bHLH-Zip transcription factor | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
51599 | |
4° C | |
Liquid |
Immunoprecipitation, Western Blot | |
1 mg/ml | |
phosphate, tris citrate with 0.09% sodium azide; pH 7-8 | |
Q86X29 | |
LSR | |
Between 450 and 500 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.