Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ LSS Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5115524
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5115524 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5115524 Supplier Invitrogen™ Supplier No. PA5115524
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total LSS.

The protein encoded by this gene catalyzes the conversion of (S)-2, 3 oxidosqualene to lanosterol. The encoded protein is a member of the terpene cyclase/mutase family and catalyzes the first step in the biosynthesis of cholesterol, steroid hormones, and vitamin D. Alternative splicing results in multiple transcript variants encoding different isoforms.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LSS
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide
Gene LSS
Gene Accession No. P48449
Gene Alias 2,3-epoxysqualene-lanosterol cyclase; 2,3-epoxysqualene--lanosterol cyclase; 2,3-oxidosqualene: lanosterol cyclase; 2810025N20Rik; CTRCT44; D10Ertd116e; hOSC; lanosterol synthase; lanosterol synthase (2,3-oxidosqualene-lanosterol cyclase); LSS; OSC; oxidosqualene lanosterol cyclase; oxidosqualene--lanosterol cyclase
Gene Symbols LSS
Host Species Rabbit
Immunogen A synthesized peptide derived from human LSS(Accession P48449), corresponding to amino acid residues A354-R404.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4047
Target Species Human, Monkey
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.