Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
LST1 Antibody (LST1/02), mFluor Violet 450 SE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP145072MFV450
Description
LST1 Monoclonal antibody specifically detects LST1 in Human samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence, ImmunoprecipitationSpecifications
LST1 | |
Monoclonal | |
mFluor Violet 450 SE | |
50mM Sodium Borate | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
Western Blot, Immunocytochemistry, Immunoprecipitation | |
LST1/02 | |
Western Blot, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation | |
B144D6S49ELST-1, leucocyte specific transcript 1, leukocyte specific transcript 1, leukocyte-specific transcript 1 protein, lymphocyte antigen 117, MGC119006, MGC119007, Protein B144 | |
KLH-fused peptide corresponding to amino acids SSEGPDLRGRDKRGT of human LST1 | |
0.1 mL | |
Cell Cycle and Replication | |
7940 | |
Store at 4C in the dark. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction