Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ LST1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5116258
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5116258 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5116258 Supplier Invitrogen™ Supplier No. PA5116258
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

LST1 Polyclonal Antibody for Western Blot

LST1 (leukocyte-specific transcript, also known as B144) is expressed in cells of myeloid/erythroid lineage (monocytes, granulocytes, dendritic cells, platelets, erythrocytes). At least 14 alternatively spliced variants (LST1/A - LST1/N) can be detected; some of them (LST1/A, B, C, G, I, K) are transmembrane cell surface-exposed forms, the other are soluble. LST1 induces production of long, thin filopodia in dendritic cells, has an inhibitory effect on lymphocyte proliferation and may have an immunomodulatory role. LST1/A is an 11 kDa transmembrane adaptor present in membrane rafts and forms spontaneously covalent homodimers. Its intracellular domain contains two tyrosine motifs, one of them being an ITIM very similar to such motifs in Siglec.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LST1
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene LST1
Gene Accession No. O00453
Gene Alias B144; D6S49E; DAAP-90L16.2; leukocyte specific transcript 1; leukocyte-specific transcript 1 protein; LST1; LST-1; lymphocyte antigen 117; MGC119006; MGC119007; Protein B144
Gene Symbols LST1
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 19-47 amino acids from the Central region of human LST1.
Purification Method Antigen affinity chromatography, Protein A
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7940
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.