Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ LTA4H Monoclonal Antibody (OTI1E9)
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA524986
Description
LTA4H Monoclonal Antibody for Western Blot, IHC (P), IP
Hydrolyzes an epoxide moiety of leukotriene A4 (LTA-4) to form leukotriene B4 (LTB-4). The enzyme also has some peptidase activity.
Specifications
LTA4H | |
Monoclonal | |
0.7 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P09960, P30349 | |
LTA4H | |
Full-length protein expressed in 293T cell transfected with human LTA4H expression vector. | |
100 μL | |
Primary | |
Human, Rat, Canine, Monkey | |
Antibody | |
IgG2b |
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
OTI1E9 | |
Unconjugated | |
LTA4H | |
Leukotriene A(4) hydrolase; leukotriene A4 hydrolase; Leukotriene A-4 hydrolase; LKHA4; LTA4; LTA4 hydrodase; LTA-4 hydrolase; LTA4H; testicular secretory protein Li 27 | |
Mouse | |
Affinity chromatography | |
RUO | |
299732, 4048, 482611 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction