Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LTA4H Mouse anti-Canine, Human, Rat, Clone: OTI1E9, lyophilized, TrueMAB™
SDP

Catalog No. 501659733
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-165-9733 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-165-9733 Supplier OriGene Supplier No. CF500666
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO

Hydrolyzes an epoxide moiety of leukotriene A4 (LTA-4) to form leukotriene B4 (LTB-4). The enzyme also has some peptidase activity.

Specifications

Antigen LTA4H
Applications Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Monoclonal
Clone OTI1E9
Conjugate Unconjugated
Formulation PBS with 8% trehalose and no preservative; pH 7.3
Gene LTA4H
Gene Accession No. P09960, P30349
Gene Alias leukotriene A4 hydrolase
Gene Symbols LTA4H
Host Species Mouse
Immunogen Full-length protein expressed in 293T cell transfected with human LTA4H expression vector
Purification Method Affinity Chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 299732, 4048, 482611
Target Species Canine, Human, Rat
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Lyophilized
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.