Learn More
LTA4H Mouse anti-Human, Clone: OTI6B5, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF500663

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
Hydrolyzes an epoxide moiety of leukotriene A4 (LTA-4) to form leukotriene B4 (LTB-4). The enzyme also has some peptidase activity.Specifications
LTA4H | |
Monoclonal | |
Unconjugated | |
LTA4H | |
leukotriene A4 hydrolase | |
Mouse | |
Affinity Chromatography | |
RUO | |
4048 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Flow Cytometry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
OTI6B5 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
P09960 | |
LTA4H | |
Full length human recombit protein of human LTA4H produced in HEK293T cell. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG2b |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.