Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ LTA4H Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA551138
Description
The antibody detects endogenous levels of total Leukotriene A4 hydrolase protein.
Hydrolyzes an epoxide moiety of leukotriene A4 (LTA-4) to form leukotriene B4 (LTB-4). The enzyme also has some peptidase activity.
Specifications
LTA4H | |
Polyclonal | |
Unconjugated | |
LTA4H | |
Leukotriene A(4) hydrolase; leukotriene A4 hydrolase; Leukotriene A-4 hydrolase; LKHA4; LTA4; LTA4 hydrodase; LTA-4 hydrolase; LTA4H; testicular secretory protein Li 27 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
16993, 4048 | |
-20°C | |
Liquid |
Immunohistochemistry, Western Blot | |
1 mg/mL | |
PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 | |
P09960, P24527 | |
LTA4H | |
Fusion protein of human LTA4H. | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction