Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ LTA4H Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA552553
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA552553 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA552553 Supplier Invitrogen™ Supplier No. PA552553
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: ALTVQSQEDN LRSLVLDTKD LTIEKVVING QEVKYALGER QSYKGSPMEI SLPIALSKNQ EIVIEISFET SPKSSALQWL TPEQTSGKEH PYLFSQCQAI HCRAILPCQD TPSVKLTYTA EVSVPKELVA LMSAIRDGET Highest antigen sequence identity to the following orthologs: Mouse - 95%, Rat - 95%.

Hydrolyzes an epoxide moiety of leukotriene A4 (LTA-4) to form leukotriene B4 (LTB-4). The enzyme also has some peptidase activity.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LTA4H
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene LTA4H
Gene Accession No. P09960
Gene Alias Leukotriene A(4) hydrolase; leukotriene A4 hydrolase; Leukotriene A-4 hydrolase; LKHA4; LTA4; LTA4 hydrodase; LTA-4 hydrolase; LTA4H; testicular secretory protein Li 27
Gene Symbols LTA4H
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human LTA4H. Recombinant protein control fragment (Product #RP-89566).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4048
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.