Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ LTA4H Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA553537
Description
Immunogen sequence: YLERHICGRL FGEKFRHFNA LGGWGELQNS VKTFGETHPF TKLVVDLTDI DPDVAYSSVP YEKGFALLFY LEQLLGGPEI FLGFLKAYVE KFSYKSITTD DWKDFLYSYF KDKVDVLNQV DWNAWLYSPG LPPIKPNYDM TL Highest antigen sequence identity to the following orthologs: Mouse - 87%, Rat - 88%.
Hydrolyzes an epoxide moiety of leukotriene A4 (LTA-4) to form leukotriene B4 (LTB-4). The enzyme also has some peptidase activity.
Specifications
| LTA4H | |
| Polyclonal | |
| Unconjugated | |
| LTA4H | |
| Leukotriene A(4) hydrolase; leukotriene A4 hydrolase; Leukotriene A-4 hydrolase; LKHA4; LTA4; LTA4 hydrodase; LTA-4 hydrolase; LTA4H; testicular secretory protein Li 27 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 4048 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| P09960 | |
| LTA4H | |
| Recombinant protein corresponding to Human LTA4H. Recombinant protein control fragment (Product #RP-89565). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction