Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ LTA4H Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA580783
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA580783 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA580783 Supplier Invitrogen™ Supplier No. PA580783
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%). This antibody has specificity for Human Leukotriene A4 Hydrolase/LTA4H.

Hydrolyzes an epoxide moiety of leukotriene A4 (LTA-4) to form leukotriene B4 (LTB-4). The enzyme also has some peptidase activity.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LTA4H
Applications ELISA, Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene LTA4H
Gene Accession No. P09960
Gene Alias Leukotriene A(4) hydrolase; leukotriene A4 hydrolase; Leukotriene A-4 hydrolase; LKHA4; LTA4; LTA4 hydrodase; LTA-4 hydrolase; LTA4H; testicular secretory protein Li 27
Gene Symbols LTA4H
Host Species Rabbit
Immunogen Recombinant Human Leukotriene A4 Hydrolase/LTA4H protein (Met1-Asp611).
Purification Method Antigen affinity chromatography, Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4048
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.