Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ LTA4H Recombinant Rabbit Monoclonal Antibody (455)
GREENER_CHOICE

Catalog No. PIMA530654
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA530654 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA530654 Supplier Invitrogen™ Supplier No. MA530654
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%). Recombinant rabbit monoclonal antibodies are produced using in vitro expression systems. The expression systems are developed by cloning in the specific antibody DNA sequences from immunoreactive rabbits. Then, individual clones are screened to select the best candidates for production. The advantages of using recombinant rabbit monoclonal antibodies include: better specificity and sensitivity, lot-to-lot consistency, animal origin-free formulations, and broader immunoreactivity to diverse targets due to larger rabbit immune repertoire. This antibody has specificity for Human Leukotriene A4 Hydrolase/LTA4H.

Hydrolyzes an epoxide moiety of leukotriene A4 (LTA-4) to form leukotriene B4 (LTB-4). The enzyme also has some peptidase activity.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LTA4H
Applications ELISA
Classification Recombinant Monoclonal
Clone 455
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene LTA4H
Gene Accession No. P09960
Gene Alias Leukotriene A(4) hydrolase; leukotriene A4 hydrolase; Leukotriene A-4 hydrolase; LKHA4; LTA4; LTA4 hydrodase; LTA-4 hydrolase; LTA4H; testicular secretory protein Li 27
Gene Symbols LTA4H
Host Species Rabbit
Immunogen Recombinant Human Leukotriene A4 Hydrolase/LTA4H protein (Met1-Asp611).
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4048
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.