Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
R&D Systems™ LTB4 Parameter Assay Kit

Click to view available options
Sufficient For:
1 x 96-well Plate
6 x 96-well Plates
Description
The LTB4 Parameter Assay Kit quantifies multi-species Leukotriene B4/LTB4 in cell culture supernates (50 ul), serum (50 ul), edta plasma (50 ul), heparin plasma (50 ul).
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
Assay Range | 10.30 to 2500 pg/mL |
Assay Sensitivity | 10.9 pg/mL |
Conjugate | Biotin |
Cross Reactivity | Cross-species reactivity not tested. <0.5% cross-reactivity observed with available related molecules. |
Product Type | ELISA Kit |
Sample Type | Cell Culture Supernatant, Plasma (EDTA), Plasma (Heparin), Serum |
Specificity | LTB4 Levels in Various Samples |
Sufficient For | 1 x 96-well Plate |
Target Species | Multi-species |
For Use With (Application) | ELISA |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction