Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ LTB4R Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA533881
Description
Percent identity with other species by BLAST analysis: Human, Gorilla, Gibbon, Marmoset (100%) Rat, Rabbit (95%) Monkey, Mouse, Hamster (90%).
LTB4R (Leukotriene B4 Receptor) is a member of the GPCR family. It is a receptor for leukotriene B4, a potent chemoattractant involved in inflammation and immune response. It has been reported in blood, bone marrow, brain, heart, liver, and skeletal muscle. In addition, it has been shown to be highly expressed in leukocytes and lymphoid tissues such as spleen, thymus, and lymph node. ESTs have been isolated from B-cell/lung/testis, breast, colon, eye, kidney, skin, spleen, testis, and tonsil libraries.
Specifications
LTB4R | |
Polyclonal | |
Unconjugated | |
LTB4R | |
BLT; BLT1; BLT-1; BLTR; Chemoattractant receptor-like 1; chemokine receptor-like 1; CMKRL1; G protein-coupled receptor 16; GPR16; G-protein coupled receptor 16; leukotriene B4 receptor; leukotriene B4 receptor 1; LTB4 receptor; Ltb4r; LTB4-R 1; Ltb4r1; LTB4-R1; LTBR1; mBLTR; P2RY7; P2Y purinoceptor 7; P2Y7; purinergic receptor P2Y, G-protein coupled, 7 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
1241 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
1 mg/mL | |
PBS with 0.1% sodium azide | |
Q15722 | |
LTB4R | |
Synthetic 20 amino acid peptide from 1st cytoplasmic domain of human Leukotriene B4 Receptor. | |
50 μg | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction