Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ LTB4R2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA533912
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA533912 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA533912 Supplier Invitrogen™ Supplier No. PA533912
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Percent identity with other species by BLAST analysis: Human, Gorilla, Monkey, Marmoset (100%) Mouse, Rat, Bat (88%) Elephant (81%).

BLTR2 has been reported in humans in peripheral blood leukocytes, mononuclear lymphocytes, and spleen. ESTs have been isolated from a broad array of human libraries, including brain, eye, fetal lung/testis/B-cell, heart, kidney, melanocyte/uterus/fetal heart, testis, tonsil, and uterus libraries.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LTB4R2
Applications Flow Cytometry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% sodium azide
Gene LTB4R2
Gene Accession No. Q9NPC1
Gene Alias 5830462O07Rik; BLT2; Blt-2; BLT2R; BLTR2; Hblt2; JULF2; KPG_004; Leukotriene B4 receptor 2; leukotriene B4 receptor BLT2; LTB4 receptor JULF2; LTB4-R 2; LTB4R2; LTB4-R2; LTB4-R-2; NOP9; Seven transmembrane receptor BLTR2
Gene Symbols LTB4R2
Host Species Rabbit
Immunogen Synthetic 16 amino acid peptide from 3rd cytoplasmic domain of human LTB4R2.
Purification Method Antigen Affinity Chromatography
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 56413
Target Species Human, Monkey
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.