Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ LTBP1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5119249
Change view
Click to view available options
Quantity:
20 μL
200 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5119249 200 μL
PIPA5121790 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. PIPA5119249 Supplier Invitrogen™ Supplier No. PA5119249
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

LTBP1 Polyclonal Antibody for Western Blot, IHC (P)

The protein encoded by this gene belongs to the family of latent TGF-beta binding proteins. The secretion and activation of TGF-betas is regulated by their association with latency-associated proteins and with latent TGF-beta binding proteins. The product of this gene targets latent complexes of transforming growth factor beta to the extracellular matrix, where the latent cytokine is subsequently activated by several different mechanisms. Alternatively spliced transcript variants encoding different isoforms have been identified.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LTBP1
Applications Immunohistochemistry (Paraffin), Western Blot, Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4
Gene LTBP1
Gene Accession No. Q8CG19
Gene Alias 9430031G15Rik; 9830146M04; b2b1000Clo; LanC (bacterial lantibiotic synthetase component C)-like 1; latent TGF beta binding protein; latent transforming growth factor beta binding protein 1; latent transforming growth factor binding protein 1; latent transforming growth factor binding protein-1; latent-transforming growth factor beta-binding protein 1; LTBP1; LTBP-1; MGC163161; Tgfb; TGF-beta-1-BP-1; TGF-beta1-BP-1; transforming growth factor beta-1-binding protein 1; Transforming growth factor beta-1-masking protein large subunit; transforming growth factor-beta (TGF-beta) masking protein large subunit
Gene Symbols LTBP1
Host Species Rabbit
Immunogen Recombinant protein Latent Transforming Growth Factor Beta Binding Protein 1. The antigen corresponds to amino acid range 1415-1712 of the target protein.
Purification Method Antigen affinity chromatography, Protein A
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 268977
Target Species Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.