Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ LTBP1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA599878
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA599878 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA599878 Supplier Invitrogen™ Supplier No. PA599878
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total LTBP1.

The protein encoded by this gene belongs to the family of latent TGF-beta binding proteins. The secretion and activation of TGF-betas is regulated by their association with latency-associated proteins and with latent TGF-beta binding proteins. The product of this gene targets latent complexes of transforming growth factor beta to the extracellular matrix, where the latent cytokine is subsequently activated by several different mechanisms. Alternatively spliced transcript variants encoding different isoforms have been identified.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LTBP1
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene LTBP1
Gene Accession No. Q00918, Q14766, Q8CG19
Gene Alias 9430031G15Rik; 9830146M04; b2b1000Clo; LanC (bacterial lantibiotic synthetase component C)-like 1; latent TGF beta binding protein; latent transforming growth factor beta binding protein 1; latent transforming growth factor binding protein 1; latent transforming growth factor binding protein-1; latent-transforming growth factor beta-binding protein 1; LTBP1; LTBP-1; MGC163161; Tgfb; TGF-beta-1-BP-1; TGF-beta1-BP-1; transforming growth factor beta-1-binding protein 1; Transforming growth factor beta-1-masking protein large subunit; transforming growth factor-beta (TGF-beta) masking protein large subunit
Gene Symbols LTBP1
Host Species Rabbit
Immunogen A synthesized peptide derived from human LTBP1(Accession Q14766), corresponding to amino acid residues S1674-E1721.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 268977, 4052, 59107
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.