Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ LTBP4 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA567624
Description
LTBP4 Polyclonal Antibody for IHC (P)
The protein encoded by this gene binds transforming growth factor beta (TGFB) as it is secreted and targeted to the extracellular matrix. TGFB is biologically latent after secretion and insertion into the extracellular matrix, and sheds TGFB and other proteins upon activation. Defects in this gene may be a cause of cutis laxa and severe pulmonary, gastrointestinal, and urinary abnormalities. Three transcript variants encoding different isoforms have been found for this gene.
Specifications
LTBP4 | |
Polyclonal | |
Unconjugated | |
LTBP4 | |
2310046A13Rik; ARCL1C; latent transforming growth factor beta binding protein 4; latent transforming growth factor-beta binding protein 4L; latent-transforming growth factor beta-binding protein 4; LTBP4; LTBP-4; LTBP4L; LTBP4S | |
Rabbit | |
Affinity Chromatography | |
RUO | |
8425 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.9 mg/mL | |
PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 | |
Q8N2S1 | |
LTBP4 | |
Synthesized peptide derived from human LTBP4. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction