Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ LTBR Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5102743
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5102743 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5102743 Supplier Invitrogen™ Supplier No. PA5102743
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total LTBR.

The protein encoded by this gene is a member of the tumor necrosis factor (TNF) family of receptors. It is expressed on the surface of most cell types, including cells of epithelial and myeloid lineages, but not on T and B lymphocytes. The protein specifically binds the lymphotoxin membrane form (a complex of lymphotoxin-alpha and lymphtoxin-beta). The encoded protein and its ligand play a role in the development and organization of lymphoid tissue and tranformed cells. Activation of the encoded protein can trigger apoptosis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LTBR
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene Ltbr
Gene Accession No. P36941, P50284
Gene Alias AI256028; CD18; D12S370; LT beta-R; Ltar; LT-beta receptor; LTbetaR; LT-BETA-R; Ltbr; lymphotoxin B receptor; lymphotoxin beta receptor; lymphotoxin beta receptor (TNFR superfamily, member 3); lymphotoxin-beta receptor; TNF receptor-related protein; Tnfbr; TNFCR; TNFR2-RP; TNFR3; TNF-RIII; TNF-R-III; TNFR-III; TNFRrp; TNFR-RP; Tnfrsf3; Tumor necrosis factor C receptor; tumor necrosis factor receptor 2-related protein; Tumor necrosis factor receptor superfamily member 3; tumor necrosis factor receptor superfamily, member 3; tumor necrosis factor receptor type III
Gene Symbols Ltbr
Host Species Rabbit
Immunogen A synthesized peptide derived from human LTBR(Accession P36941), corresponding to amino acid residues L24-R74.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 17000, 297604, 4055
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.