Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ LTBR Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA579613
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA579613 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA579613 Supplier Invitrogen™ Supplier No. PA579613
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 μg/mL. Positive Control - WB: HELA whole cell, A549 whole cell. IHC: Human Intestinal Cancer Tissue.

The protein encoded by this gene is a member of the tumor necrosis factor (TNF) family of receptors. It is expressed on the surface of most cell types, including cells of epithelial and myeloid lineages, but not on T and B lymphocytes. The protein specifically binds the lymphotoxin membrane form (a complex of lymphotoxin-alpha and lymphtoxin-beta). The encoded protein and its ligand play a role in the development and organization of lymphoid tissue and tranformed cells. Activation of the encoded protein can trigger apoptosis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LTBR
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 500 μg/mL
Conjugate Unconjugated
Formulation PBS with 5mg BSA and 0.05mg sodium azide, 0.05mg thimerosal
Gene Ltbr
Gene Accession No. P36941
Gene Alias AI256028; CD18; D12S370; LT beta-R; Ltar; LT-beta receptor; LTbetaR; LT-BETA-R; Ltbr; lymphotoxin B receptor; lymphotoxin beta receptor; lymphotoxin beta receptor (TNFR superfamily, member 3); lymphotoxin-beta receptor; TNF receptor-related protein; Tnfbr; TNFCR; TNFR2-RP; TNFR3; TNF-RIII; TNF-R-III; TNFR-III; TNFRrp; TNFR-RP; Tnfrsf3; Tumor necrosis factor C receptor; tumor necrosis factor receptor 2-related protein; Tumor necrosis factor receptor superfamily member 3; tumor necrosis factor receptor superfamily, member 3; tumor necrosis factor receptor type III
Gene Symbols Ltbr
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence at the C-terminus of human LTBR (420-435aa ATPSNRGPRNQFITHD).
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4055
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Lyophilized
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.