Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LTBR Polyclonal Antibody, Invitrogen™

Catalog No. PIPA588290
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA588290 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA588290 Supplier Thermo Scientific Supplier No. PA588290

Rabbit Polyclonal Antibody

Immunogen sequence: KSHPSLCRKL GSLLKRRPQG EGPNPVAGSW EPPKAHPYFP DLVQPLLPIS GDVSPVSTGL PAAPVLEAGV PQQQSPLDLT REPQLEPGEQ SQVAHGTNGI HVTGGSMTIT GNIYIYNGPV LGGPPGPGDL PATPEPPYPI PEEGDPGPPG LSTPHQEDGK AWHLAETEHC GATPSNRGPR NQFITHD; Positive Samples: Mouse liver; Cellular Location: Membrane, Single-pass type I membrane protein

The protein encoded by this gene is a member of the tumor necrosis factor (TNF) family of receptors. It is expressed on the surface of most cell types, including cells of epithelial and myeloid lineages, but not on T and B lymphocytes. The protein specifically binds the lymphotoxin membrane form (a complex of lymphotoxin-alpha and lymphtoxin-beta). The encoded protein and its ligand play a role in the development and organization of lymphoid tissue and tranformed cells. Activation of the encoded protein can trigger apoptosis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LTBR
Applications Western Blot
Classification Polyclonal
Concentration 0.94 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal, pH 7.3
Gene Ltbr
Gene Accession No. P50284
Gene Alias AI256028, CD18, D12S370, LT beta-R, Ltar, LT-beta receptor, LTbetaR, LT-BETA-R, Ltbr, lymphotoxin B receptor, lymphotoxin beta receptor, lymphotoxin beta receptor (TNFR superfamily, member 3), lymphotoxin-beta receptor, TNF receptor-related protein, Tnfbr, TNFCR, TNFR2-RP, TNFR3, TNF-RIII, TNF-R-III, TNFR-III, TNFRrp, TNFR-RP, Tnfrsf3, Tumor necrosis factor C receptor, tumor necrosis factor receptor 2-related protein, Tumor necrosis factor receptor superfamily member 3, tumor necrosis factor receptor superfamily, member 3, tumor necrosis factor receptor type III
Gene Symbols Ltbr
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 249-435 of human LTBR (NP_0023331)
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 17000
Target Species Mouse
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.