Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ltbr, Rat anti-Mouse, Clone: 3C8, Abnova™

Catalog No. 89998882 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-998-882 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-998-882 Supplier Abnova Corporation Supplier No. MAB16107
Add to Cart
Add to Cart

Rat Monoclonal Antibody

Specifications

Antigen Ltbr
Applications Functional Assay
Classification Monoclonal
Clone 3C8
Conjugate Unconjugated
Dilution Functional Study (agonist inducing BAFF, chemokines and integrins in vitro and in vivo) The optimal working dilution should be determined by the end user.
Formulation In PBS (10% glycerol, 0.02% sodium azide).
Gene lymphotoxin B receptor
Gene Alias AI256028/CD18/LTbetaR/Ltar/TNF-R-III/TNFCR/TNFR-RP/TNFR2-RP/TNFRrp/Tnfbr/Tnfrsf3
Gene Symbols Ltbr
Host Species Rat
Immunogen Recombinant protein corresponding to amino acids 31-221 of mouse Ltbr.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 17000
Target Species Mouse
Content And Storage For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.