Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Ltbr, Rat anti-Mouse, Clone: 3C8, Abnova™
Rat Monoclonal Antibody
Supplier: Abnova Corporation MAB16108
Specifications
Ltbr | |
Monoclonal | |
Unconjugated | |
In PBS. | |
AI256028/CD18/LTbetaR/Ltar/TNF-R-III/TNFCR/TNFR-RP/TNFR2-RP/TNFRrp/Tnfbr/Tnfrsf3 | |
Rat | |
100 μg | |
Primary | |
Mouse | |
Antibody | |
IgG1 κ |
Functional Assay | |
3C8 | |
Functional Study (agonist inducing BAFF, chemokines and integrins in vitro and in vivo) The optimal working dilution should be determined by the end user. | |
lymphotoxin B receptor | |
Ltbr | |
Recombinant protein corresponding to amino acids 31-221 of mouse Ltbr. | |
RUO | |
17000 | |
For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction