Learn More
Invitrogen™ LTC4S Polyclonal Antibody

Description
LTC4S Polyclonal Antibody for Western Blot
Specifications
Specifications
| Antigen | LTC4S |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.09% sodium azide; pH 7.4 |
| Gene | LTC4S |
| Gene Accession No. | Q16873 |
| Gene Alias | Leukotriene C4 synthase; Leukotriene-C(4) synthase; LTC4 synthase; LTC4S |
| Gene Symbols | LTC4S |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.