Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ LTK Polyclonal Antibody
GREENER_CHOICE

Catalog No. PA130314
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PA130314 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PA130314 Supplier Invitrogen™ Supplier No. PA130314
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive Control: Placenta lysate.

LTK (TYK1), ALK and Ros have been identified as receptor tyrosine kinases having sequence similarity to the Insulin receptor subfamily of kinases. LTK, leukocyte tyrosine kinase, is expressed in murine B-lymphocyte precursors and has also been found in forebrain neurons. ALK, anaplastic lymphoma kinase, is normally highly expressed specifically in the nervous system. A truncated form containing the catalytic domain of ALK is expressed as the result of a translocation occurring in many non-Hodgkin's lymphomas. The c-Ros gene was originally identified in mutant form as an oncogene. Ros is normally expressed in a small number of epithelial cell types and may play a role in epithelial development.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LTK
Applications Western Blot
Classification Polyclonal
Concentration 1.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide
Gene LTK
Gene Accession No. P29376
Gene Alias EC 2.7.1.112; EC 2.7.10.1; leukocyte receptor tyrosine kinase; leukocyte tyrosine kinase; leukocyte tyrosine kinase receptor; Ltk; Protein tyrosine kinase 1; TYK1; tyrosine kinase-1
Gene Symbols LTK
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 42-72 amino acids from the N-terminal region of human LTK.
Purification Method Ammonium sulfate precipitation
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4058
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.