Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LTRIN Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB10178110
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB10178110 100 μg
MAB101781SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. MAB10178110 Supplier R&D Systems Supplier No. MAB101781100
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

LTRIN Monoclonal specifically detects LTRIN in Multi-Species samples. It is validated for Western Blot, ELISA.

Specifications

Antigen LTRIN
Applications Western Blot, ELISA
Classification Monoclonal
Clone 1009809
Conjugate Unconjugated
Dilution Western Blot 2 ug/mL, ELISA
Formulation Lyophilized from a 0.2 µm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 µm filtered solution in PBS.
Host Species Mouse
Immunogen Synthetic Aedes aegypti LTRIN peptide
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23687673
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Multi-species
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.