Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ LTV1 Homolog Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA556372
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA556372 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA556372 Supplier Invitrogen™ Supplier No. PA556372
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: ATGEEEGMDI QKSENEDDSE WEDVDDEKGD SNDDYDSAGL LSDEDCMSVP GKTHRAIADH LFWSEETKSR FTEYSM Highest antigen sequence identity to the following orthologs: Mouse - 55%, Rat - 49%.

LTV1 gene ontology annotations related to this gene include ribosomal small subunit biogenesis; ribosomal small subunit export from nucleus.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LTV1 Homolog
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.8 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene LTV1
Gene Accession No. Q96GA3
Gene Alias 2610020N02Rik; C6orf93; dJ468K18.4; Ltv1; LTV1 homolog; LTV1 homolog (S. cerevisiae); LTV1 ribosome biogenesis factor; protein LTV1 homolog
Gene Symbols LTV1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human LTV1 Homolog. Recombinant protein control fragment (Product #RP-94841).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84946
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.