Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ LU AA33810
GREENER_CHOICESDP

Catalog No. 356210R Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
35-621-0R 10 mg
35-625-0R 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 35-621-0R Supplier Tocris Bioscience™ Supplier No. 3562/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

Potent NPY Y5 receptor antagonist

LU AA33810 is a potent neuropeptide Y (NPY) Y5 receptor antagonist (Ki = 1.5 nM in vitro). Displays ≥ 3300-fold affinity for Y5 over Y1, Y2 and Y4 receptors. Also binds human 5-HT2B and 5-HT1A receptors (Ki values are 247 and 478 nM respectively). Exerts anxiolytic- and antidepressant-like effects in rat models of stress sensitivity.

Specifications

Quantity 10 mg
Product Type LU AA33810
Purity >98%
Molecular Weight (g/mol) 423.62
Inhibitors LU AA33810

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.