Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lubricin/SZP Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501981143
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-1143 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-1143 Supplier Bioss Supplier No. BS11175R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Lubricin is a glycoprotein encoded by the PRG4 gene. It has been identified as a component of joint lubrication and synovial homeostasis by reducing friction in joints and preventing synovial cell overgrowth. It is a large, water soluble molecule that acts as a carrier for insoluble surface-active phospholipid (SAPL). Depletion of lubricin function has been associated with camptodactyly-arthropathy-coxa vara-pericarditis syndrome (CACP), an arthritic-like autosomal recessive disorder.

Specifications

Antigen Lubricin/SZP
Applications Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene PRG4
Gene Accession No. Q92954
Gene Alias articular superficial zone protein; CACP; HAPO; hemangiopoietin; JCAP; Lubricin; megakaryocyte stimulating factor; Megakaryocyte-stimulating factor; MSF; PRG4; Proteoglycan 4; Proteoglycan 4 C-terminal part; Proteoglycan-4; Superficial zone proteoglycan; SZP
Gene Symbols PRG4
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human Lubricin/SZP.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10216
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.