Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Luc-Screen™ Extended-Glow Luciferase Reporter Gene Assay System

Catalog No. T1035
Encompass_Preferred
Click to view available options
Quantity:
10,000 Assays
1000 Assays
200 Assays

Includes

Two reaction buffers

Thermo Scientific Luc-Screen Extended-Glow Luciferase Reporter Gene Assay System with extended-glow light emission is designed for sensitive detection of firefly luciferase reporter enzyme in microplate cell cultures. A linear signal is obtained with the

  • Homogeneous assay format allows detection of firefly luciferase in presence of normal culture media without removal of media and without an additional cell lysis step, providing easiest and most streamlined assay procedure possible
  • Glow assay kinetics provides a window during which measurements may be performed, facilitating HTS applications where assay automation is used
  • Wide dynamic range of firefly luciferase assay lets the user measure enzyme level accurately from femtogram to nanogram range
  • Assay sensitivity is 100- to 1,000-fold better than with either the isotopic/non-isotopic assays for chloramphenicol acetyl transferase (CAT) or the colorimetric/fluorescent assays for β-galactosidase, providing greater sensitivity than competing assay technologies
  • Highly sensitive assay with a wide dynamic range permits detection of high and low levels of reporter without performing numerous sample dilutions
  • Non-radioactive reporter gene assay kit eliminates radioisotopes
  • Assay can be completed in about one hour, providing fast assay turnaround

Sensitive
Luciferase is an ideal reporter because of the high sensitivity of detection and the absence of endogenous luciferase activity in mammalian cells. The Luc-Screen assay conveniently couples in-well cell lysis in the presence of culture medium with a high sensitivity assay that exhibits extended-glow light emission kinetics. Light signal can be measured between 10 minutes and several hours after adding assay reagents, providing great flexibility in the time between reagent addition and measurement.

High Throughput
The Luc-Screen system is designed for maximum assay flexibility in a high throughput format and can be used in luminometers without automatic injectors. The Luc-Screen system is formulated to provide a convenient, easy-to-use luciferase assay that is optimized for use in high-throughput screening. The high sensitivity of the Luc-Screen assay is accompanied by a wide dynamic range.

Specifications

  • Assay Type: Luciferase Assay
  • Compatible Cells: Mammalian Cells
  • Detection Method: Chemiluminescent
  • For Use With Equipment: Luminometer (Microplate)
  • Form: Liquid
  • Format: 96-well plate
  • Readout: End Point
  • Sample Type (Specific): Cell Cultures
  • Substrate: Luciferin
  • Substrate Properties: Chemical Substrate
  • Substrate Type: Luciferase Substrate
TRUSTED_SUSTAINABILITY

Specifications

Product Type Luciferase Reporter Gene Assay System
Detection Method Chemiluminescence
For Use With (Equipment) Luminometer (Microplate)
Assay Luciferase Assay
Content And Storage This kit contains two reaction buffers.
Store the product at -20°C.
Compatible Cells Mammalian Cells
High-throughput Compatibility High-throughput Compatible
Readout End Point
Sample Type Cell Cultures
Substrate Luciferin
Substrate Type Luciferase Substrate
Substrate Properties Chemical Substrate
Target Enzyme Luciferase
Product Line Luc-Screen™, NovaBright™
For Use With (Application) Enzyme Assay
Quantity 200 Assays
Form Liquid
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.