Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
LUC7L Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP179695
Description
LUC7L Polyclonal specifically detects LUC7L in Human samples. It is validated for Western Blot.Specifications
LUC7L | |
Polyclonal | |
Unconjugated | |
PBS, 2% Sucrose with 0.09% Sodium Azide | |
FLJ10231, hLuc7B1, Luc7, LUC7 (S. cerevisiae)-like, LUC7B1, LUC7L1, LUC7-LIKE, LUC7-like (S. cerevisiae), putative RNA-binding protein Luc7-like 1, Putative SR protein LUC7B1, sarcoplasmic reticulum protein LUC7B1, SR+89 | |
Rabbit | |
36 kDa | |
100 μL | |
Primary | |
Expected identity based on immunogen sequence: Bovine: 100%; Chicken: 100%; Canine: 100%; Equine: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Xenopus: 92%; Guinea pig: 92%; Zebrafish: 92%. | |
Human, Mouse, Rat, Bovine, Canine, Equine, Guinea Pig, Rabbit, Zebrafish | |
IgG |
Western Blot | |
0.5 mg/ml | |
Western Blot 1.0 ug/ml | |
NP_060502 | |
LUC7L | |
Synthetic peptide directed towards the middle region of human LUC7LThe immunogen for this antibody is LUC7L. Peptide sequence EEIGKLLAKAEQLGAEGNVDESQKILMEVEKVRAKKKEAEEEYRNSMPAS. | |
Affinity purified | |
RUO | |
55692 | |
Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction