Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ LUC7L Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA566014
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA566014 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA566014 Supplier Invitrogen™ Supplier No. PA566014
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: GHRRASRDRSA KYKFSRERAS REESWES Highest antigen sequence identity to the following orthologs - mouse 86%, rat 86%.

May bind to RNA via its Arg/Ser-rich domain.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LUC7L
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Luc7l
Gene Accession No. Q9NQ29
Gene Alias 1810045C04Rik; 2410018D03Rik; hLuc7B1; Luc7; Luc7 homolog (S. cerevisiae)-like; LUC7 like; LUC7B1; Luc7l; LUC7L1; LUC7-like; LUC7-like (S. cerevisiae); Putative RNA-binding protein Luc7-like 1; putative SR protein LUC7B1; sarcoplasmic reticulum protein LUC7B1; SR+89
Gene Symbols Luc7l
Host Species Rabbit
Immunogen Recombinant Human LUC7L. Recombinant protein control fragment (Product #RP-105041).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55692
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.