Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LUC7L3 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501983187
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-3187 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-3187 Supplier Bioss Supplier No. BS14067R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Binds cAMP regulatory element DNA sequence. May play a role in RNA splicing.

Specifications

Antigen LUC7L3
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene LUC7L3
Gene Accession No. O95232
Gene Alias 3300001P08Rik; BOS_18986; cAMP regulatory element-associated protein 1; cisplatin resistance associated overexpressed protein; cisplatin resistance-associated overexpressed protein; Cisplatin resistance-associated-overexpressed protein; CRA; CREAP1; CREAP-1; CRE-associated protein 1; Crop; FLJ11063; hLuc7A; LOC100340089; LUC7 like 3 pre-mRNA splicing factor; Luc7A; Luc7l3; LUC7-like 3; LUC7-like 3 (S. cerevisiae); LUC7-like 3 pre-mRNA splicing factor; Luc7-like protein 3; O48; OA48-18; Okadaic acid-inducible phosphoprotein OA48-18; RGD1307981; zgc:136380; zgc:158165
Gene Symbols LUC7L3
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human LUC7L3.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51747
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.