Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Vidan™ Diagnostics Lucidplus™ Beta-hydroxybutyrate (BHB) Starter Package
Click to view available options
:
Lucidplus™ Beta-hydroxybutyrate (BHB) Starter Package
Description
- 1 Lucidplus™ Monitoring System
- 1 kit of Lucidplus™ BHB Test Strips (50 tests)
- 1 box of Lucidplus™ BHB B-level Controls (2 x 4 ml)
- Drop ship from Vidan
Specifications
Specifications
Includes | (1) Lucidplus Montoring System, (1) Lucidplus BHB Test Strips, (1) Lucidplus Bi-level Controls |
Depth (Metric) | 13.5 cm |
Height (Metric) | 11.0 cm |
Width (Metric) | 19.5 cm |
Weight (Metric) | 0.45 kg |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction