Learn More
Invitrogen™ Lucifer Yellow Polyclonal Antibody, Biotin

Description
Lucifer Yellow Polyclonal Antibody for IHC, ICC/IF
Specifications
Specifications
| Antigen | Lucifer Yellow |
| Applications | Immunohistochemistry, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 3 mg/mL |
| Conjugate | Biotin |
| Formulation | PBS with 5mM sodium azide; pH 7.2 |
| Gene Alias | Lucifer Yellow Dye |
| Host Species | Rabbit |
| Immunogen | Lucifer Yellow. |
| Purification Method | IgG fraction |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.