Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
                
Learn More
Learn More
                                    Luciferase Antibody (Luci 21 1-107), DyLight 650, Novus Biologicals™
 
                                    
                                    
                                    
                                    
                                
                            
                            
                            
                            
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB600307C
Description
Luciferase Monoclonal specifically detects Luciferase in Firefly samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.Specifications
| Luciferase | |
| Monoclonal | |
| DyLight 650 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Firefly | |
| Purified | 
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Luci 21 1-107 | |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen | |
| ec 1.13.12.7, Fluc, luciferin 4 monooxygenase | |
| This Luciferase Antibody (Luci 21 1-107) was developed against luciferase protein from Photinus pyralis (North American firefly). [UniProt# P08659]. | |
| 0.1 mL | |
| Cell Biology, Cellular Markers, Epitope Tags, Fusion Proteins | |
| This Luciferase Antibody (Luci 21 1-107) is specific for Luciferase, recognizing a peptide consisting of the first 258 amino acids. Further epitope mapping has not been done at this time. | |
| Store at 4C in the dark. | |
| IgG1 κ | 
                    Product Content Correction
                
                Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
			
            For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction
            