Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Luciferase Antibody (Luci 21 1-107), mFluor Violet 500 SE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB600307MFV500
Description
Luciferase Monoclonal antibody specifically detects Luciferase in Firefly samples. It is validated for Flow CytometrySpecifications
Luciferase | |
Monoclonal | |
mFluor Violet 500 SE | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified | |
RUO | |
Firefly | |
Purified |
Flow Cytometry | |
Luci 21 1-107 | |
Flow Cytometry | |
ec 1.13.12.7, firefly luciferase, Fluc, LuC, luciferin 4 monooxygenase, Luciferin 4-monooxygenase | |
This Luciferase Antibody (Luci 21 1-107) was developed against luciferase protein from Photinus pyralis (North American firefly). [UniProt# P08659]. | |
0.1 mL | |
Primary | |
Store at 4C in the dark. | |
IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction