Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Luciferase Antibody, Novus Biologicals™

Goat Polyclonal Antibody has been used in 19 publications
Supplier: Novus Biologicals NB1001677100UG
Description
Luciferase Polyclonal specifically detects Luciferase in Firefly samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.Specifications
Luciferase | |
Polyclonal | |
Unconjugated | |
0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2 with 0.01% Sodium Azide | |
ec 1.13.12.7, Fluc, luciferin 4 monooxygenase | |
Native protein from Photinus pyralis (Firefly). | |
100 μg | |
Primary | |
Firefly | |
Purified |
Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
1.0 mg/mL | |
Western Blot 1:1000 - 1:5000, ELISA 1:200 - 1:1000, Immunohistochemistry 1:1000-1:5000, Immunocytochemistry/Immunofluorescence 20 ug/ml, Immunohistochemistry-Paraffin 1:1000 - 1:5000, Immunohistochemistry-Frozen 1:1000 - 1:5000 | |
P08659 | |
Goat | |
Ion exchange chromatography | |
RUO | |
Luciferase, no reactivity is observed against Sea pansy (Renilla reniformis) luciferase. | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction