Learn More
Luciferase Polyclonal Antibody, HRP, Rockland™

Goat Polyclonal Antibody
$178.35 - $578.10
Specifications
| Antigen | Luciferase |
|---|---|
| Concentration | 1 mg/mL |
| Applications | ELISA, Immunoprecipitation, Western Blot |
| Classification | Polyclonal |
| Conjugate | HRP |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-105-7523
![]()
|
Rockland Immunochemicals
200103150S |
25 μL |
Each for $178.35
|
|
|||||
50-199-0348
![]()
|
Rockland Immunochemicals
2001031500100 |
100 μL |
Each for $578.10
|
|
|||||
Description
Store vial at 4°C prior to restoration. For extended storage aliquot contents and freeze at -20°C or below. Avoid cycles of freezing and thawing. Centrifuge product if not completely clear after standing at room temperature. This product is stable for several weeks at 4°C as an undiluted liquid. Dilute only prior to immediate use. Assay by immunoelectrophoresis resulted in a single precipitin arc against anti-Peroxidase, anti-Goat Serum as well as purified and partially purified Luciferase [Photinus pyralis (Firefly)]. No reactivity is observed against Sea pansy (Renilla reniformis) luciferase.
Luciferase produces green light with the wavelength of 562 nm. Luciferase catalyzes the reaction: D-firefly luciferin + O2 + ATP = firefly oxyluciferin + CO2 + AMP + diphosphate + light.Specifications
| Luciferase | |
| ELISA, Immunoprecipitation, Western Blot | |
| HRP | |
| Goat | |
| Firefly | |
| Firefly Luciferase; Genji firefly; Green Luciferase; Japanese firefly; Photinus pyralis | |
| Primary | |
| Antibody |
| 1 mg/mL | |
| Polyclonal | |
| Lyophilized | |
| RUO | |
| 0.02M potassium phosphate with 0.15M NaCl, 10 mg/mL BSA and 0.01% gentamicin sulfate; pH 7.2 | |
| Luciferase [Photinus pyralis (Firefly)] | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.