Learn More
Description
- Reagent for Detection of Pro-Luciferin Substrates
- Reagent available without esterase
- Use in combination with the pro-luciferin substrates
Specifications
Specifications
| Quantity | 50 mL |
| Content And Storage | 50 mL luciferin detection reagent and 50 mL reconstitution buffer. -30°C to -10°C |
| For Use With (Application) | Monitor the activity of specific isoforms of cytochrome P450 or NAT2 |
| pH | 7.2 |
| Product Type | Luciferin |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.