Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ LUF 5834
GREENER_CHOICESDP

Catalog No. 460350 Shop All Novus Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
46-035-0 50 mg
46-031-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 46-035-0 Supplier R&D Systems™ Supplier No. 4603/50

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Potent adenosine A2A and A2B receptor partial agonist

LUF 5834 is a potent A2A and A2B adenosine receptor partial agonist (Ki = 2.6 nM and EC50 = 12 nM respectively). Also exhibits selectivity for A1 over A3 (Ki values are 2.6 and 538 nM respectively).

Chemical Identifiers

CAS 333962-91-7
Molecular Formula C17H12N6OS

Specifications

Quantity 50 mg
Product Line Tocris
Formula Weight 348.38
Molecular Weight (g/mol) 348.384
Chemical Name or Material LUF 5834
Recommended Storage Store at -20°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.