Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lugol's Dilute Solution, 2%
SCI_ED

Catalog No. s25945 Shop All Fisherbrand Products
Click to view available options
Quantity:
500 mL
Packaging:
2%
This item is not returnable. View return policy
This item is not returnable. View return policy

Specifications

Chemical Name or Material Lugol's Dilute Solution, 2%
Color Brown
CAS 7681-11-0, 7553-56-2, 7732-18-5
Physical Form Liquid
Quantity 500 mL
Synonym Tincture of Iodine
Grade Laboratory
Packaging 2%
Density 1 to 1.05
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.