Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ LUM Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA597941
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA597941 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA597941 Supplier Invitrogen™ Supplier No. PA597941
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

LUM Polyclonal Antibody for Western Blot, ELISA

LUM is a member of the small leucine-rich proteoglycan (SLRP) family that includes decorin, biglycan, fibromodulin, keratocan, epiphycan, and osteoglycin. In these bifunctional molecules, this protein moiety binds collagen fibrils and the highly charged hydrophilic glycosaminoglycans regulate interfibrillar spacings. The protein is the major keratan sulfate proteoglycan of the cornea but is also distributed in interstitial collagenous matrices throughout the body. Lumican may regulate collagen fibril organization and circumferential growth, corneal transparency, and epithelial cell migration and tissue repair.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LUM
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 2.62 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene LUM
Gene Accession No. P51886
Gene Alias keratan sulfate proteoglycan lumican; KSPG lumican; Lcn; Ldc; Lum; Lumican; Lumican precursor; lumican proteoglycan; SLRR2D
Gene Symbols LUM
Host Species Rabbit
Immunogen Recombinant Rat Lumican protein (19-338AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 81682
Target Species Rat
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.