Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ LUM Recombinant Rabbit Monoclonal Antibody (JE11-45)
GREENER_CHOICE

Catalog No. PIMA534828
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA534828 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA534828 Supplier Invitrogen™ Supplier No. MA534828
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

Positive Control: Jurkat, 293T, Human skin tissue, MCF-7, MG-63, PANC-1, rat kidney tissue, human skin tissue.

LUM is a member of the small leucine-rich proteoglycan (SLRP) family that includes decorin, biglycan, fibromodulin, keratocan, epiphycan, and osteoglycin. In these bifunctional molecules, this protein moiety binds collagen fibrils and the highly charged hydrophilic glycosaminoglycans regulate interfibrillar spacings. The protein is the major keratan sulfate proteoglycan of the cornea but is also distributed in interstitial collagenous matrices throughout the body. Lumican may regulate collagen fibril organization and circumferential growth, corneal transparency, and epithelial cell migration and tissue repair.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LUM
Applications Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone JE11-45
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.05% BSA, 40% glycerol and 0.05% sodium azide; pH 7.4
Gene LUM
Gene Accession No. P51884
Gene Alias keratan sulfate proteoglycan lumican; KSPG lumican; Lcn; Ldc; Lum; Lumican; Lumican precursor; lumican proteoglycan; SLRR2D
Gene Symbols LUM
Host Species Rabbit
Immunogen Recombinant protein within Human Lumican aa 172-338.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4060
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles, store in dark
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.