Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Lumican Antibody (110), Janelia Fluor™ 549 , Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP289942JF549
Description
Lumican Monoclonal antibody specifically detects Lumican in Human samples. It is validated for ELISASpecifications
Lumican | |
Monoclonal | |
Janelia Fluor 549 | |
KSPG lumican, lumican, lumican proteoglycan, SLRR2DLDCKeratan sulfate proteoglycan lumican | |
This antibody was obtained from a rabbit immunized with purified, recombinant Human Lumican (NP_002336.1; Met1-Asn338). | |
0.1 mL | |
Cancer, Cell Biology, Cytoskeleton Markers, Signal Transduction | |
4060 | |
Store at 4°C in the dark. | |
IgG |
ELISA | |
110 | |
50mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction